Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079693.4 | NP_524417.1 | 1 - 2 | 236 | 69 | |
GenBank mRNA | AY061281.1 | - | 1 - 2 | 243 | 69 | |
U28966.1 | - | 1 - 2 | 232 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |