Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260283.2 | NP_001247212.1 | 7 - 8 | 2814 | 68 | |
NM_001260284.2 | NP_001247213.1 | 6 - 7 | 2669 | 68 | ||
NM_001260285.2 | NP_001247214.1 | 6 - 7 | 2621 | 68 | ||
NM_001260286.2 | NP_001247215.1 | 7 - 8 | 3532 | 68 | ||
NM_001300505.1 | NP_001287434.1 | 6 - 7 | 2774 | 68 | ||
NM_001300506.1 | NP_001287435.1 | 6 - 7 | 4188 | 68 | ||
NM_079699.4 | NP_524423.2 | 5 - 6 | 3463 | 68 | ||
NM_169940.4 | NP_732576.1 | 6 - 7 | 2866 | 68 | ||
NM_169941.2 | NP_732577.1 | 5 - 6 | 2708 | 68 | ||
GenBank mRNA | AY118680.1 | - | 6 - 7 | 2812 | 68 | |
BT011420.1 | - | 6 - 7 | 2809 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |