Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc93AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275864.1 | NP_001262793.1 | 16 - 17 | 8535 | 64 | |
NM_079700.3 | NP_524424.2 | 15 - 16 | 8505 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |