Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc93AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275864.1 | NP_001262793.1 | 24 - 25 | 11705 | 78 | |
NM_079700.3 | NP_524424.2 | 23 - 24 | 11675 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |