Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170197.1 | NP_001163668.1 | 6 - 7 | 3196 | 92 | |
NM_001260292.1 | NP_001247221.1 | 7 - 8 | 1080 | 92 | ||
NM_001275866.1 | NP_001262795.1 | 7 - 8 | 1067 | 92 | ||
NM_079701.5 | NP_524425.2 | 7 - 8 | 1383 | 92 | ||
NM_169944.3 | NP_732584.1 | 7 - 8 | 1067 | 92 | ||
NM_206529.2 | NP_996251.2 | 7 - 8 | 1383 | 92 | ||
GenBank mRNA | AY058543.1 | - | 7 - 8 | 1342 | 92 | |
BT044567.1 | - | 6 - 7 | 3194 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |