Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079705.3 | NP_524429.1 | 4 - 5 | 1199 | 62 | |
GenBank mRNA | AF181656.1 | - | 4 - 5 | 1151 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |