Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275872.1 | NP_001262801.1 | 5 - 6 | 1680 | 70 | |
NM_001275873.1 | NP_001262802.1 | 5 - 6 | 1680 | 70 | ||
NM_079706.3 | NP_524430.1 | 5 - 6 | 1680 | 70 | ||
GenBank mRNA | AF222923.1 | - | 5 - 6 | 1677 | 70 | |
AF222924.1 | - | 5 - 6 | 1559 | 70 | ||
AY118898.1 | - | 5 - 6 | 1648 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |