Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170210.2 | NP_001163681.1 | 4 - 5 | 782 | 85 | |
NM_001275882.1 | NP_001262811.1 | 4 - 5 | 782 | 85 | ||
NM_079717.5 | NP_524441.2 | 4 - 5 | 782 | 85 | ||
GenBank mRNA | BT046172.1 | - | 4 - 5 | 766 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |