Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079718.3 | NP_524442.2 | 1 - 2 | 1263 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |