Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300530.1 | NP_001287459.1 | 2 - 3 | 521 | 60 | |
NM_079722.4 | NP_524446.3 | 2 - 3 | 521 | 60 | ||
NM_169991.2 | NP_732694.2 | 3 - 4 | 575 | 60 | ||
GenBank mRNA | AY071402.1 | - | 3 - 4 | 575 | 60 | |
AY119620.1 | - | 2 - 3 | 497 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |