Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079730.3 | NP_524454.2 | 3 - 4 | 1171 | 113 | |
GenBank mRNA | AY060363.1 | - | 3 - 4 | 1153 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |