Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beta-Man | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259970.2 | NP_001246899.1 | 2 - 3 | 246 | 71 | |
NM_001275312.1 | NP_001262241.1 | 1 - 2 | 359 | 71 | ||
NM_001275313.1 | NP_001262242.1 | 1 - 2 | 359 | 71 | ||
NM_141179.4 | NP_649436.2 | 1 - 2 | 359 | 71 | ||
GenBank mRNA | BT044157.1 | - | 1 - 2 | 258 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |