Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275320.1 | NP_001262249.1 | 2 - 3 | 654 | 73 | |
NM_001275321.1 | NP_001262250.1 | 2 - 3 | 654 | 73 | ||
NM_141196.2 | NP_649453.1 | 2 - 3 | 654 | 73 | ||
GenBank mRNA | AY122072.1 | - | 2 - 3 | 323 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |