Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043211.2 | NP_001036676.1 | 4 - 5 | 1731 | 79 | |
NM_001104215.2 | NP_001097685.2 | 4 - 5 | 1731 | 79 | ||
NM_141248.4 | NP_649505.1 | 4 - 5 | 1731 | 79 | ||
NM_169022.2 | NP_730882.2 | 4 - 5 | 1731 | 79 | ||
GenBank mRNA | AY060484.1 | - | 4 - 5 | 1709 | 79 | |
BT024186.1 | - | 3 - 4 | 721 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |