Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043211.2 | NP_001036676.1 | 5 - 6 | 1966 | 67 | |
NM_001104215.2 | NP_001097685.2 | 5 - 6 | 1966 | 67 | ||
NM_141248.4 | NP_649505.1 | 5 - 6 | 1966 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |