Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275355.1 | NP_001262284.1 | 6 - 7 | 2142 | 67 | |
NM_001300246.1 | NP_001287175.1 | 6 - 7 | 2304 | 67 | ||
NM_001300247.1 | NP_001287176.1 | 6 - 7 | 2160 | 67 | ||
NM_141285.2 | NP_649542.1 | 6 - 7 | 2286 | 67 | ||
GenBank mRNA | AY118365.1 | - | 6 - 7 | 2283 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |