Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA1-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144539.2 | NP_001138011.1 | 6 - 7 | 2218 | 84 | |
NM_001275364.2 | NP_001262293.1 | 8 - 9 | 1873 | 84 | ||
NM_141309.4 | NP_649566.2 | 7 - 8 | 1765 | 84 | ||
NM_169086.3 | NP_730976.1 | 8 - 9 | 1873 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |