Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xe7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141315.4 | NP_649572.4 | 3 - 4 | 1430 | 74 | |
NM_169093.3 | NP_730984.1 | 3 - 4 | 1430 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |