Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141354.3 | NP_649611.2 | 4 - 5 | 782 | 66 | |
GenBank mRNA | AY069336.1 | - | 4 - 5 | 750 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |