Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300296.1 | NP_001287225.1 | 3 - 4 | 549 | 69 | |
NM_001300297.1 | NP_001287226.1 | 3 - 4 | 549 | 69 | ||
NM_141512.3 | NP_649769.1 | 3 - 4 | 549 | 69 | ||
GenBank mRNA | AB185208.1 | - | 3 - 4 | 434 | 69 | |
AY094809.1 | - | 3 - 4 | 502 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |