Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043221.4 | NP_001036686.3 | 3 - 4 | 333 | 68 | |
NM_001300302.1 | NP_001287231.1 | 2 - 3 | 210 | 68 | ||
NM_141518.4 | NP_649775.2 | 3 - 4 | 379 | 68 | ||
NM_169212.3 | NP_731221.2 | 4 - 5 | 499 | 68 | ||
GenBank mRNA | BT044586.1 | - | 1 - 2 | 136 | 68 | |
BT126358.1 | - | 2 - 3 | 194 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |