Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170094.2 | NP_001163565.1 | 1 - 2 | 211 | 70 | |
NM_001275476.1 | NP_001262405.1 | 1 - 2 | 211 | 70 | ||
NM_141611.2 | NP_649868.1 | 1 - 2 | 211 | 70 | ||
GenBank mRNA | BT150443.1 | - | 1 - 2 | 161 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |