Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141611.2 | NP_649868.1 | 7 - 8 | 3914 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |