Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141637.3 | NP_649894.1 | 1 - 2 | 380 | 66 | |
GenBank mRNA | AY061527.1 | - | 1 - 2 | 355 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |