Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P58IPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141659.4 | NP_649916.1 | 2 - 3 | 631 | 74 | |
GenBank mRNA | AY058567.1 | - | 2 - 3 | 616 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |