Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bruce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275531.2 | NP_001262460.1 | 9 - 10 | 4505 | 93 | |
NM_001275532.2 | NP_001262461.1 | 8 - 9 | 4205 | 93 | ||
NM_001275533.2 | NP_001262462.1 | 8 - 9 | 4205 | 93 | ||
NM_001275534.2 | NP_001262463.1 | 8 - 9 | 4205 | 93 | ||
NM_141738.5 | NP_649995.2 | 8 - 9 | 4205 | 93 | ||
GenBank mRNA | AF517634.1 | - | 8 - 9 | 4202 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |