Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104268.2 | NP_001097738.2 | 3 - 4 | 1206 | 73 | |
NM_001260092.2 | NP_001247021.1 | 3 - 4 | 1206 | 73 | ||
NM_001260094.2 | NP_001247023.2 | 3 - 4 | 1230 | 73 | ||
NM_141742.3 | NP_649999.2 | 3 - 4 | 1230 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |