Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141760.4 | NP_650017.1 | 3 - 4 | 621 | 96 | |
GenBank mRNA | AY095041.1 | - | 3 - 4 | 582 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |