Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141863.3 | NP_650120.1 | 2 - 3 | 512 | 82 | |
NM_169434.2 | NP_731639.1 | 2 - 3 | 437 | 82 | ||
NM_169435.3 | NP_731640.1 | 2 - 3 | 512 | 82 | ||
GenBank mRNA | AY089465.1 | - | 2 - 3 | 594 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |