Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316533.1 | NP_001303462.1 | 3 - 4 | 669 | 61 | |
NM_141879.3 | NP_650136.1 | 3 - 4 | 669 | 61 | ||
GenBank mRNA | AY058745.1 | - | 3 - 4 | 624 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |