Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275565.1 | NP_001262494.1 | 2 - 3 | 1052 | 73 | |
NM_141886.3 | NP_650143.1 | 2 - 3 | 1052 | 73 | ||
NM_169446.2 | NP_731664.1 | 1 - 2 | 1150 | 73 | ||
GenBank mRNA | AY051950.1 | - | 2 - 3 | 1033 | 73 | |
BT015287.1 | - | 1 - 2 | 1117 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |