Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260146.2 | NP_001247075.1 | 10 - 11 | 6211 | 61 | |
NM_141936.2 | NP_650193.1 | 10 - 11 | 6205 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |