Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141983.3 | NP_650240.1 | 4 - 5 | 1563 | 60 | |
NM_169482.1 | NP_731749.3 | 5 - 6 | 1840 | 60 | ||
GenBank mRNA | AF218811.1 | - | 4 - 5 | 1532 | 60 | |
BT100045.1 | - | 4 - 5 | 1495 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |