Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheA87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142023.4 | NP_650280.1 | 1 - 2 | 252 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |