Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Droj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142026.3 | NP_650283.1 | 2 - 3 | 1268 | 69 | |
NM_169519.2 | NP_731804.1 | 2 - 3 | 1406 | 69 | ||
NM_169520.2 | NP_731805.1 | 2 - 3 | 1195 | 69 | ||
NM_169521.2 | NP_731806.1 | 1 - 2 | 1245 | 69 | ||
NM_169522.2 | NP_731807.1 | 3 - 4 | 1209 | 69 | ||
GenBank mRNA | AY060982.1 | - | 2 - 3 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |