Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142032.3 | NP_650289.2 | 2 - 3 | 320 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |