Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142033.2 | NP_650290.2 | 3 - 4 | 1881 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |