Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144572.2 | NP_001138044.1 | 8 - 9 | 3344 | 104 | |
NM_142034.2 | NP_650291.1 | 9 - 10 | 3464 | 104 | ||
GenBank mRNA | BT015265.1 | - | 9 - 10 | 3454 | 104 | |
EF078694.1 | - | 8 - 9 | 2632 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |