Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Task6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275610.1 | NP_001262539.1 | 3 - 4 | 1399 | 90 | |
NM_001275611.1 | NP_001262540.1 | 3 - 4 | 2447 | 90 | ||
NM_001275612.1 | NP_001262541.1 | 3 - 4 | 2447 | 90 | ||
NM_142043.3 | NP_650300.2 | 3 - 4 | 1399 | 90 | ||
GenBank mRNA | AJ920330.1 | - | 2 - 3 | 287 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |