Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142070.3 | NP_650327.1 | 1 - 2 | 587 | 91 | |
GenBank mRNA | AY118299.1 | - | 1 - 2 | 572 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |