Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142074.4 | NP_650331.1 | 2 - 3 | 184 | 54 | |
NM_169568.3 | NP_731880.1 | 2 - 3 | 219 | 54 | ||
GenBank mRNA | AY084159.1 | - | 2 - 3 | 178 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |