Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275637.2 | NP_001262566.1 | 8 - 9 | 2780 | 59 | |
NM_001300398.1 | NP_001287327.1 | 8 - 9 | 2421 | 59 | ||
NM_142112.5 | NP_650369.3 | 8 - 9 | 3034 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |