Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR-C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142144.3 | NP_650401.1 | 2 - 3 | 127 | 91 | |
NM_169607.2 | NP_731954.1 | 1 - 2 | 199 | 91 | ||
GenBank mRNA | AY058243.1 | - | 1 - 2 | 80 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |