Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyc88E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043246.4 | NP_001036711.1 | 6 - 7 | 1112 | 109 | |
NM_001260186.2 | NP_001247115.1 | 5 - 6 | 1018 | 109 | ||
NM_169616.4 | NP_731974.4 | 5 - 6 | 1018 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |