Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275659.1 | NP_001262588.1 | 2 - 3 | 265 | 84 | |
NM_142185.2 | NP_650442.1 | 2 - 3 | 265 | 84 | ||
GenBank mRNA | AF152245.1 | - | 2 - 3 | 250 | 84 | |
AY051679.1 | - | 2 - 3 | 241 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |