Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202307.2 | NP_001189236.1 | 3 - 4 | 311 | 68 | |
NM_001202308.2 | NP_001189237.1 | 3 - 4 | 296 | 68 | ||
NM_142315.4 | NP_650572.2 | 3 - 4 | 252 | 68 | ||
NM_169727.2 | NP_732167.1 | 2 - 3 | 390 | 68 | ||
GenBank mRNA | AY089543.1 | - | 3 - 4 | 220 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |