Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abd-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275719.2 | NP_001262648.1 | 6 - 7 | 1347 | 76 | |
NM_001316542.1 | NP_001303471.1 | 6 - 7 | 6271 | 76 | ||
NM_001316543.1 | NP_001303472.1 | 4 - 5 | 2502 | 76 | ||
NM_001316544.1 | NP_001303473.1 | 6 - 7 | 1347 | 76 | ||
NM_001316545.1 | NP_001303474.1 | 6 - 7 | 1347 | 76 | ||
NM_080157.3 | NP_524896.2 | 4 - 5 | 2502 | 76 | ||
NM_142320.2 | NP_650577.1 | 6 - 7 | 1165 | 76 | ||
NM_169735.2 | NP_732180.1 | 6 - 7 | 1338 | 76 | ||
NM_169736.2 | NP_732181.1 | 6 - 7 | 1347 | 76 | ||
NM_206498.2 | NP_996220.1 | 6 - 7 | 1333 | 76 | ||
GenBank mRNA | BT124750.1 | - | 6 - 7 | 1343 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |