Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142337.3 | NP_650594.1 | 2 - 3 | 840 | 107 | |
NM_169744.3 | NP_732202.2 | 2 - 3 | 602 | 107 | ||
NM_176508.3 | NP_788685.1 | 2 - 3 | 390 | 107 | ||
GenBank mRNA | BT099945.1 | - | 2 - 3 | 598 | 107 | |
DQ372684.1 | - | 2 - 3 | 466 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |