Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170164.2 | NP_001163635.2 | 4 - 5 | 1341 | 53 | |
NM_142356.3 | NP_650613.2 | 3 - 4 | 960 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |