Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TyrRII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275753.1 | NP_001262682.1 | 4 - 5 | 785 | 75 | |
NM_001275754.2 | NP_001262683.1 | 4 - 5 | 1423 | 75 | ||
NM_001300451.1 | NP_001287380.1 | 4 - 5 | 1423 | 75 | ||
NM_142394.2 | NP_650651.1 | 3 - 4 | 986 | 75 | ||
GenBank mRNA | BT029098.1 | - | 3 - 4 | 849 | 75 | |
DQ151546.1 | - | 3 - 4 | 986 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |